Learn More
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -111.0°C |
Density | 1.2620g/mL |
Boiling Point | 46.0°C |
Quantity | 100 mL |
Flash Point | −30°C |
Packaging | Glass Bottle |
Refractive Index | 1.627 |
Assay Percent Range | 99.85% min. (GC) |
Specific Gravity | 1.262 |
Linear Formula | CS2 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465292 | Certificate of Analysis | 12/08/2024 | 445660010,445661000,445660025 |
A0465293 | Certificate of Analysis | 07/08/2024 | 445660010,445661000,445660025 |
A0462822 | Certificate of Analysis | 14/05/2024 | 445660010,445661000,445660025 |
A0443815 | Certificate of Analysis | 10/01/2024 | 445660010,445661000,445660025 |
A0458176 | Certificate of Analysis | 30/11/2023 | 445660010,445661000,445660025 |
Safety and Handling



- Carbon disulfide
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.