Learn More
Erlab™ Captair by Erlab™ H402 Version 03 Filtration Columns
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ H402 Version 03 Filtration Columns are for storing organic vapors and acids
Brand: Erlab™ G0103/F
This item is not returnable.
View return policy
Description
- Horizontal filtration column
- HEPA H14 filter
- Handlings in an ultra clean environment
- Serial prefilter(optimizes the HEPA H14 filter lifetime)
- Optional carbon filtration = clean the air polluted by ambient air VOCs
Specifications
Filtration Column | |
For storing Formaldehyde |
ERL030 Catalogue global-2011-FR-chemtrap
File size: 1.6 MB
ERL030 Page chemtrap GB
File size: 558.9 kB
Product Content Correction
Product Title
Erlab™ Captair by Erlab™ H402 Version 03 Filtration Columns > Filter Type: F Carbon