Learn More
Thermo Scientific™ Caps for Nunclon™ Delta Flasks
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.
Brand: Thermo Scientific™ 133001
94.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close (solid) and filter—to meet your application needs.
Vent/close Caps- EasY Flask vent/close caps feature a "Y" mark that allows visual verification
- Vent position easily distinguised even what flasks are stacked in incubators
- Filter membrane provides effective barrier against microorganisms
- Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
- Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
- Sterilized by gamma irradiation and certified to be nonpyrogenic
- Case of 100
Specifications
Blue | |
HDPE | |
100 | |
Vent/close cap | |
Solid | |
Nunclon Delta |
Sterile, HDPE closure | |
175 cm² EasY Flask | |
1 | |
Sterile | |
175 cm2, 225 cm2 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
179668 | Certificate of Compliance | 02/12/2024 | 133001 |
172220 | Certificate of Compliance | 02/12/2024 | 133001 |
165213 | Certificate of Compliance | 02/12/2024 | 133001 |
177456 | Certificate of Compliance | 02/12/2024 | 133001 |
172220 | Certificate of Compliance | 23/05/2022 | 133001 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.