Learn More
Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kit
Collect, store and transport urine samples for drugs of abuse testing with Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kits.
Brand: Thermo Scientific™ 90111
122.67 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kits provide a collection beaker, transport vial, and specimen transport bag in a tamper evident package to support drug testing programs. The kits are constructed from a low-binding polypropropylene resin that has been laboratory tested for interaction with drugs of abuse analytes.
- Easy-to-use—simply peel away the foil-top lid, remove the components and provide donor with the collection cup
- After specimen is collected, transfer urine into the tamper-evident vial, close the hinged lid until it snaps, and bag for transport
Ordering Information:
Case of 100. Custom branding available for foil tops and specimen bags.
Includes:
Collection cup (8 oz.) with temperature indicator strip, 45mL Tamper evident transport vial, dual pocket specimen bag with absorbent pad, and a foil top seal
Specifications
Specimen Collection Kit | |
Sterile | |
Single Specimen Collection Kit |
Polypropylene | |
45 mL | |
Collection cup (8 oz.) with temperature indicator strip, 45 mL Tamper evident transport vial, Generic dual pocket specimen bag, Absorbent pad and Foil top |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
New Offer Save Now
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.