Learn More
Fisherbrand™ Capillary Tube
Accessory for use with 2D Vertical Gel Electrophoresis Systems.
Brand:  Fisherbrand™ MCT10
Product Code. 11877653
  • 41.00€ / Pack of 10
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Measures 75mm L with 1mm I.D.
Specifications
Specifications
2D Vertical Gel Electrophoresis System
Capillary Tube
75 mm
1 mm
Documents
One moment while we fetch your results.
Product Title
Fisherbrand™ Capillary Tube >