Learn More
Unity Lab Services™ Calibration Certificate Cofrac Temperature Two Fixed Points, 0°C, 120°C
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand:  Unity Lab Services™ UMV-P935A
Product Code. 12655670
  • 85.00€ / Each
Explore more special offers
Description
Description
  • Calibration by comparison with a reference thermometer in baths
  • Type of device: temperature measuring thermometric chain or expansion thermometer, at fixed values of 0°C, 120°C
  • Our best uncertainties for a thermometer with a resolution of 0.01°C (k = 2) +/- 0.02°C at 0°C; +/- 0.08°C to 120°C
  • Our best uncertainties for a thermometer with a resolution of 0.1°C (k = 2) +/- 0.1°C to 0°C; +/- 0.2°C to 120°C
  • COFRAC Accreditation Number 2-5984
  • Calibration, scope available on www.cofrac.fr
  • Average time: 1 week
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermometers and Temperature Recorders
Cofrac
Calibration Certificate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Unity Lab Services™ Calibration Certificate Cofrac Temperature Two Fixed Points, 0°C, 120°C >