Learn More
Manutan Cable ties, 200mm x 4.8mm
Brand:  Manutan A000174
Encompass_Preferred
Product Code. 12815609
  • POA / Pack of 1000
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Tie and fix quickly and securely. UL-proved nylon 6/6. Weather and waterproof, temperature resistance from -35 to +85°C.
Dimensions L x W : 200mm x 4.8mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Manutan Cable ties, 200mm x 4.8mm