Learn More
VACUUBRAND™ Cable For VV-B and VV-B/C In-line Isolation Valves
Brand: VACUUBRAND™ 20612556
104.50 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Specifications
For VV-B and VV-B/C In-line Isolation Valves | |
VACUU·BUS™ Plug | |
Cable |
200 cm | |
1 Each |
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Product Content Correction
Product Title
VACUUBRAND™ Cable For VV-B and VV-B/C In-line Isolation Valves > For use with: For VV-B and VV-B/C In-line Isolation Valves