Learn More
IKA™ C-MAG HS 7 Ceramic Plate Magnetic Stirrer Package
GREENER_CHOICE
Includes an electronic contact thermometer, support rod, boss head clamp, and holding rod in addition to the C-MAG HS 7 magnetic stirrer. IKA™ C-MAG HS 7 Ceramic Plate Magnetic Stirrer Package includes all the parts necessary for heated magnetic stirring in one convenient kit.
Brand:  IKA™ 0009015900
818.80 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10110900
  • 1380.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
Magnetic stirrer with ceramic heating plate. Bushing according to DIN 12878 for connecting a contact thermometer.
Package consists of:
  • C-MAG HS 7
  • ETS-D5 Electronic Contact Thermometer
  • H 16 V Support rod
  • H 44 Boss head clamp
  • H 38 Holding rod
C-Mag HS 7 Ceramic Plate Magnetic Stirrer offers:
  • Ceramic glass plate, offering excellent chemical resistance
  • Fixed safety circuit of 550°C
  • Hot surface warning indicator to prevent burns
  • Stirs volumes up to 10 liters (H2O)
  • Digital display
  • Digital error display
  • Elevated control panel for protection against leaking liquids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 1500 rpm
Ceramic
1020 w
180 mm
50/60 Hz
C-MAG HS 7
Magnetic Stirrer Package
1
100/120/230 V
105 mm
180 mm
C-Mag HS7, ETS-D5 contact thermometer, H16V support rod HS7, H16V rod holder, H38 holding rod, H44 boss head
50°C to 500°C
5 kg
Promotions
  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119
    Expires2025-04-30
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    16
    Hours
    0
    Minutes
    53
    Seconds
    11
Product Content Correction
Product Title
IKA™ C-MAG HS 7 Ceramic Plate Magnetic Stirrer Package > Capacity: 10L