missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH9.23 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical
Click to view available options
Quantity:
1 L
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
MDL Number | 217672 |
Packaging | Plastic container |
Safety and Handling
Product Identifier
- Buffer solution BS pH 9.23 (borate)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution pH9.23 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction