missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
Description
Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 10, MDL Number: 163147, Physical Form: Liquid
Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
- Standard and color-coded buffer solutions
- Manufactured to a tolerance of ±0.02 pH units
- Accurately standardized and labelled with lot-specific pH at 20°C
- All buffers are traceable to NIST and BS 1647 standard
- Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
- Standard buffers are available over the range pH2 to pH10
- Color-coded buffer solutions are available over the range pH4, pH7, and pH

Specifications
Specifications
Color | Blue |
pH | 10 |
Physical Form | Liquid |
Quantity | 5 L |
MDL Number | 163147 |
Packaging | Plastic container |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 3.5 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Promotions
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction