missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ea.
Specifications
Specifications
Color | Yellow |
Physical Form | Liquid |
Quantity | 6 Ea. |
Packaging | Ampoule |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 3.5 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528161 | Certificate of Analysis | 03/03/2025 | J/2855/15,J/2855C/90 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction