Learn More
Bromodeoxyuridine/BrdU Antibody (SPM166), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34744JF549
Description
Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Nonspecies specific samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
Bromodeoxyuridine/BrdU | |
Monoclonal | |
Janelia Fluor 549 | |
Bromodeoxyuridine, BUdr | |
Bromodeoxyuridine/BrdU conjugated to KLH | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Non-species specific | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
SPM166 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.