Learn More
Invitrogen™ Brilliant Stain Buffer
For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension. Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample.
142.00€ - 668.00€
Specifications
Chemical Name or Material Staining Buffer
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17460683
View Documents
Invitrogen™
00-4409-42
100 Tests
142.00€
100 tests
17470683
View Documents
Invitrogen™
00-4409-75
1000 Tests
668.00€
1000 tests
Description
Description
  • For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension
  • Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample
  • Prevents non-specific polymer interactions that can result in data appearing under-compensated
  • Buffer is provided in a 50 μL/test format
  • Compatible with traditional fluorochromes, Brilliant Violet dyes, Brilliant Ultra Violet dyes, Super Bright dyes, and standard flow cytometry protocols
  • Buffer is not compatible with UltraComp eBeads and UltraComp eBeads Plus compensation beads
  • If using these beads as a compensation tool, Flow Cytometry Staining Buffer should be used exclusively for all dilutions and washes during staining of the microspheres
Specifications
Specifications
Staining Buffer
Documents
Product Certifications
Product Content Correction
Product Title
Invitrogen™ Brilliant Stain Buffer