Learn More
Invitrogen™ Brilliant Stain Buffer
For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension. Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample.
142.00€ - 668.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17460683
|
Invitrogen™
00-4409-42 |
100 Tests |
142.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17470683
|
Invitrogen™
00-4409-75 |
1000 Tests |
668.00€
1000 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension
- Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample
- Prevents non-specific polymer interactions that can result in data appearing under-compensated
- Buffer is provided in a 50 μL/test format
- Compatible with traditional fluorochromes, Brilliant Violet dyes, Brilliant Ultra Violet dyes, Super Bright dyes, and standard flow cytometry protocols
- Buffer is not compatible with UltraComp eBeads and UltraComp eBeads Plus compensation beads
- If using these beads as a compensation tool, Flow Cytometry Staining Buffer should be used exclusively for all dilutions and washes during staining of the microspheres
Specifications
Staining Buffer |
Product Title
Invitrogen™ Brilliant Stain Buffer