Learn More
Greiner Bio-One BREATHseal™ Sealer
For the culture of prokaryotic and eukaryotic cells
Brand:  Greiner Bio-One 676050
 View more versions of this product
Product Code. 11910667
  • 4635.00€ / Pack of 50
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique gas-permeable sealer with a wide distribution of pores for excellent gas exchange
  • Available sterile and non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Translucent
Non-sterile
BREATHseal Sealer
0.14 mm
Cell Culture Microplate, HTS Microplate
Rayon-Fibers Foil, Acrylate Adhesive Glue, Poly-coated Kraft Paper with Silicone
500
Acrylic
Yes