Learn More
Alpaco™ Brass Clamps with Vertical Screws
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Brass Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
26.80€ - 29.50€
Specifications
Quantity 10/Pk.
Material Brass
View More Specs
Products 13
Product Code Brand Size (Metric) Grip Price Quantity & Availability  
Product Code Brand Size (Metric) Grip Price Quantity & Availability  
11934947
View Documents
Alpaco™
M-NH 25/10-1167
20 to 25 mm
27.60€
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11974937
View Documents
Alpaco™
5 to 10 mm
27.10€
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11914947
View Documents
Alpaco™
14 to 19 mm
27.60€
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11984937
View Documents
Alpaco™
1 to 12 mm
26.80€
Pack of 10
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
11904957
View Documents
Alpaco™
M-NH 21/10-1163
16 to 21 mm
27.60€
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11924947
View Documents
Alpaco™
M-NH 22 /10-1164
17 to 22 mm
27.60€
Pack of 10
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
11944947
View Documents
Alpaco™
M-NH 27/10-1169
22 to 27 mm
28.65€
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11954947
View Documents
Alpaco™
M-NH 30/10-1172
25 to 30 mm
28.65€
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11964947
View Documents
Alpaco™
M-NH 33/10-1175
28 to 33 mm
28.65€
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11984947
View Documents
Alpaco™
M-NH 38/10-1180
33 to 38 mm
29.50€
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11994947
View Documents
Alpaco™
M-NH 41/10-1183
36 to 41 mm
29.50€
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11904947
View Documents
Alpaco™
M-NH 17/10-1159
12 to 17 mm
27.10€
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11994937
View Documents
Alpaco™
M-NH 14/10-1156
9 to 12 mm
26.80€
Pack of 10
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
Description
Description
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifications
Specifications
10/Pk.
Brass
Documents
Product Certifications
Product Content Correction
Product Title
Alpaco™ Brass Clamps with Vertical Screws