missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ inertGrade™ 96-well Microplates
Especially suited for cell cultures when adhesion is not desired.Reduction of cell adhesion and protein adsorption minimizes enzyme and cellular activation to aid in the inhibition of the differentiation of progenitor or stem cells and promotes the formation of spheroids.
148.00€ - 150.00€
Specifications
Material | Polystyrene |
---|---|
Sterility | Sterile |
Well Shape | Flat Bottom (F-well) |
No. per Pack | 5 |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
15624017
|
BRAND™
781912 |
White |
150.00€
Pack of 5 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
15634017
|
BRAND™
781913 |
Black |
148.00€
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
Description
- Hydrogel technology optimizes the surface characteristics to suppress interactions of cells or protein with the polystyrene plastic.
- Polystyrene construction
- Sterile
- 96 wells
- Flat or Round Bottom
- Black, White or Clear
- Includes lid
Specifications
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ inertGrade™ 96-well Microplates