Learn More
Borate Buffer, pH 9.00, Chem-Lab
For use in laboratory applications and pH measurements, including protein modification procedures. Borate Buffer, pH 9.00, Chem-Lab is an alkaline buffer solution traceable to Standard Reference Material (SRM) from NIST. It is comprised of Boric acid, Potassium chloride and Sodium hydroxide.
Brand: Chem Lab CL03.0218.1000
Description
A borate pH Buffer solution helps to maintain alkaline pH level via a boric acid-borate anion equilibrium. It is used in cell culture and analytical chemistry applications, including protein modification procedures.Specifications
0°C | |
Colorless | |
Laboratory use, pH measurements, traceable to SRM from NIST | |
Liquid | |
Borate (H3BO3), Potassium chloride (KCl), Sodium hydroxide (NaOH) | |
Yes |
100°C | |
1.01g/mL | |
9.00 ±0.02 | |
1 L | |
Odorless | |
Soluble in water |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.