Learn More
Bohlender™ BOLA™ Discs Stirrer Shafts for NW 200
Discs stirrer shafts include a fluoroplastic-jacketed stainless steel shaft.
Brand:  Bohlender™ C598-56
 View more versions of this product
Product Code. 17167154
  • 1823.20€ / Each
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Description
  • PTFE-jacketed stainless steel shaft
  • Discoidal stirrer blade with six radial paddles completely made of PTFE
  • Similar to a 'Rushton Turbine' stirrer shaft
  • Universal chemical resistance since product is only exposed to PTFE
Specifications
Specifications
0.39 in.
1.4 in.
36 mm
1000 mm
0.16 in.
Chucking Diameter: 8 mm
7.1 in.
PTFE
10 mm
NW 200
39 in.
45 mm
4 mm
Chucking Diameter: 8 mm
180 mm