Learn More
Brady™ Blank Vinyl Cloth Marker Cards
Brand:  Brady™ 012353
 View more versions of this product
Product Code. 15208304
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Specifications
Wire Marker Cards - Solid NEMA Colours
Polymer-coated
Matte
Marker Card
900/Pk.
0.055 mm
38 mm
Rubber, Repositionable
Black
Vinyl Cloth
Wire and cable marking
-40°C to +80°C
6.35 mm