Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Brand: Axygen™ HT-1000-CBK-HTR-5
280.33 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- Available in non-sterile, sterile, nonfiltered, and filtered
- Available as Clear or Conductive black polypropylene tips
- Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
1000 μL | |
Non-sterile | |
Not Autoclavable | |
Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
Automation | |
Hamilton MICROLAB Nimbus |
Non-filtered | |
Black | |
DNase, RNase and Pyrogen free | |
Hanging Tip Rack | |
5 Racks/Pk, 8 Pks/Cs. | |
Polypropylene |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
13821029 | Certificate of Quality | 19/12/2023 | HT-1000-CBK-HTR-5 |
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
4Hours
9Minutes
54Seconds
6