missing translation for 'onlineSavingsMsg'
Learn More
Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals Product Code.: 17582026

Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals

Product Code. 17582026
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 17582026

Brand: Thermo Scientific Alfa Aesar 1685714

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Bis(cyclopentadienyl)cobalt is used to catalyze the preparation of pyridines from alkynes and nitriles. It acts as a polymerization inhibitor of olefins. It is a common one-electron reducing agent in the laboratory. It reacts with carbon monoxide to prepare cobalt(I) derivative, cyclopentadienylcobalt dicarbonyl. It also serves as an internal standard in cyclic voltametry. Further, it finds use in Diels-Alder reactions. In addition to this, it is used as a paint drier and as an oxygen stripping agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1277-43-6
Molecular Formula C10H10Co
Molecular Weight (g/mol) 189.123
MDL Number MFCD00013749
InChI Key SNBRJOIYNQIWSZ-UHFFFAOYSA-N
Synonym Cobaltocene
PubChem CID 24942376
IUPAC Name cobalt(2+);cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]

Specifications

Melting Point 176°C to 180°C (decomposition)
Quantity 25 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 189.12
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)cobalt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY28J008 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number16857-14,016857.06,016857.03,016857.14
Lot NumberM30G015 Certificate TypeCertificate of Analysis Date23/12/2022 Catalog Number16857-14,016857.06,016857.03,016857.14
Lot NumberW30B001 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number16857-14,016857.06,016857.03,016857.14
Lot NumberH19T004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number16857-14,016857.06,016857.03,016857.14
Lot NumberQ01C019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number16857-14,016857.06,016857.03,016857.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.