missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(diisopropylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals

Product Code. 15475776
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis[2-(diisopropylphosphino)ethyl]amine acts as ligand and form complexes with ruthenium, which finds application in the dehydrogenation of ethanol.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with oxidizing agents and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131890-26-1
Molecular Formula C16H37NP2
Molecular Weight (g/mol) 305.427
MDL Number MFCD17018778
InChI Key FTVIGQGOGIHMBS-UHFFFAOYSA-N
Synonym bis 2-di-i-propylphosphino ethyl amine, di 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphanyl ethyl amine, bis 2-diisopropylphosphino ethyl amine solution, 10 wt. % in thf
PubChem CID 15757160
IUPAC Name 2-di(propan-2-yl)phosphanyl-N-[2-di(propan-2-yl)phosphanylethyl]ethanamine
SMILES CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C

Specifications

Density 0.884
Linear Formula ([(CH3)2CH]2PCH2CH2)2NH
Refractive Index 1.418
Quantity 25 g
UN Number UN2924
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 305.42
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material Bis[2-(diisopropylphosphino)ethyl]amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis[2-(diisopropylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.