missing translation for 'onlineSavingsMsg'
Learn More
Biotin, Serva Electrophoresis™ Product Code.: 11781867

Biotin, Serva Electrophoresis™

Product Code. 11781867
2.5 g
Encompass_Preferred
Click to view available options
Quantity:
2.5 g
This item is not returnable. View return policy

Product Code. 11781867

Brand: Serva Electrophoresis 1506003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 58-85-5
Molecular Formula C10H16N2O3S
MDL Number MFCD00005541
Quantity 2.5 g
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Molecular Weight (g/mol) 244.31
PubChem CID 171548
ChEBI CHEBI:15956
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Biotin, Serva Electrophoresis™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.