Description
- Lightweight low-linting CleanTough material
- ESD properties
- PPE Cat 3 Type PB [6]
- Silicone free
- Certifications and Testing:
- Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425
- Module D certificationsConforms to Category 3 - Personal Protective Equipment Regulation(EU)2016/425
- Tested against standard EN ISO 13034:2005+A1:2009 protective clothing against liquid chemicals (Type PB[6])
- Tested against standard EN ISO 14325 protective clothing against chemicals
- Tested against standards: EN 1149-5:2008, protective clothing -electrostatic properties; EN 530, Abrasion resistance; EN 7854, Flexcracking resistance; EN 6530, Resistance to penetration by liquids; EN13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN863, Puncture resistance; and EN 13935-2, Seam strength.
- Tested against permeation standard ASTM F739-12
- Processed in an NEBB certified ISO Class 4 environment
Specifications
Specifications
Product Type | Hood |
Size | Universal |
Material | Spunbond Polypropylene |
Color | White |
Closure Type | Elastic |
Disposable | Single-use |
Design Type | Three-piece |
Model | BDHD |
Seam Type | Bound, Single Needle Stitching |
Sterility | Non-sterile |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
25-0434 | Certificate of Compliance | 01/04/2025 | BDHD-L |
24-3335 | Certificate of Compliance | 04/03/2025 | BDHD-L |
24-2069 | Certificate of Compliance | 18/02/2025 | BDHD-L |
24-1706 | Certificate of Compliance | 18/02/2025 | BDHD-L |
24-1206 | Certificate of Compliance | 18/02/2025 | BDHD-L |
Product Content Correction
Product Title
Ansell BioClean-D Long-Length Non-Sterile Hood >