Learn More
Angelantoni Industrie™ Bioban Microbiological Safety Cabinets
Routinely used in laboratory of microbiology, virology, hematology and cell cultures.
Brand:  Angelantoni Industrie™ ST-003034000
Encompass
Product Code. 13585850
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Working chamber of stainless steel AISI 304L 2B finish
  • Side walls made of tempered safety glass
  • Fluorescent lamp 36w
  • Double inlet centrifugal type fan with external rotor motor and degree of protection IP 55
  • Recirculated air filtration and expulsion carried out with HEPA H14 filters
Compliance
EN12469
Specifications
Specifications
EN 12469
99.995%
Microbiology, Virology, Hematology and Cell Cultures
BIOBAN 48
HEPA H14 Filters
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Angelantoni Industrie™ Bioban Microbiological Safety Cabinets