Learn More
Tocris Bioscience™ BIBP 3226 trifluoroacetate
Mixed NPY Y1 and NPFF receptor antagonist
Brand: Tocris Bioscience™ 2707/2
This item is not returnable.
View return policy
Description
BIBP 3226 trifluoroacetate is a mixed non-peptide neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist (Ki values are 1.1, 79, 108, > 1000, > 1000 and >1000 for rNPY Y1, hNPFF2, rNPFF, rNPY Y2, rNPY Y4 and rNPY Y5 respectively). Produces an anxiogenic-like effect in rats following i.c.v. administration.
Specifications
N-[(1R)]-4-[(Aminoiminomethyl)amino-1-[[[(4-hydroxyphenyl)methyl]amino]carbonyl]butyl-α-phenylbenzeneacetamide trifluoroacetate | |
NPY Receptor Antagonists | |
98% |
2 mg | |
C27H31N5O3.xCF3CO2H |
Product Content Correction
Product Title
Tocris Bioscience™ BIBP 3226 trifluoroacetate > 2 mg