missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIBO 3304 trifluoroacetate
Highly selective NPY Y1 receptor antagonist
440.00€ - 1735.00€
Specifications
Chemical Name or Material | N-[(1R)-1-[[[[4-[[(Aminocarbonyl)amino]methyl]phenyl]methyl]amino]carbonyl]-4-[(aminoiminomethyl)amino]butyl]-α-phenyl-benzeneacetamide ditrifluoroacetate |
---|---|
CAS | 2310085-85-7 |
Assay Percent Range | >98% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C29H35N7O3.2CF3CO2H |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18005592
|
Tocris Bioscience™
2412/10 |
10 mg |
440.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18085042
|
Tocris Bioscience™
2412/50 |
50 mg |
1735.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
High affinity NPY Y1 receptor antagonist (IC50 values are 0.38 and 0.72 nM at human and rat receptors respectively) that displays > 2600-fold selectivity over Y2, Y4 and Y5 receptors. Inhibits NPY- and fasting-induced feeding in vivo following central administration. Also antagonizes anxiolytic-like effects of NPY.Specifications
N-[(1R)-1-[[[[4-[[(Aminocarbonyl)amino]methyl]phenyl]methyl]amino]carbonyl]-4-[(aminoiminomethyl)amino]butyl]-α-phenyl-benzeneacetamide ditrifluoroacetate | |
>98% | |
C29H35N7O3.2CF3CO2H |
2310085-85-7 | |
Soluble to 100mM in DMSO | |
Observed MW: 757.69 |
Safety and Handling
Recommended Storage : Store at 4°C
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title