Learn More
Tocris Bioscience™ BIBO 3304 trifluoroacetate
Highly selective NPY Y1 receptor antagonist
Brand:  Tocris Bioscience™ 2412/10
 View more versions of this product
Product Code. 18005592
  • 440.00€ / 10mg
Explore more special offers
Description
Description
BIBO 3304 trifluoroacetate is a high affinity NPY Y1 receptor antagonist (IC50 values are 0.38 and 0.72 nM at human and rat receptors respectively) that displays > 2600-fold selectivity over Y2, Y4 and Y5 receptors. Inhibits NPY - and fasting-induced feeding in vivo following central administration. Also antagonizes anxiolytic-like effects of NPY.
Specifications
Specifications
N-[(1R)-1-[[[[4-[[(Aminocarbonyl)amino]methyl]phenyl]methyl]amino]carbonyl]-4-[(aminoiminomethyl)amino]butyl]-α-phenyl-benzeneacetamide ditrifluoroacetate
>98%
Soluble to 100mM in DMSO
Observed MW: 757.69
2310085-85-7
10 mg
C29H35N7O3.2CF3CO2H