Learn More
Benzaldehyde, 99+%, Thermo Scientific Chemicals Product Code.: 11434143
Benzaldehyde, 99+%, Thermo Scientific Chemicals
Product Code. 11434143
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
Product Code. 11434143
Brand: Thermo Scientific Alfa Aesar A10348.0B
Request Bulk or Custom Format
Benzaldehyde is commonly employed as a precursor to prepare various organic compounds, which are used as pharmaceuticals, dyes, perfumes, flavorings, photographic chemicals and plastic additives. It is used as solvent for oils and resins. Benzaldehyde is further used as a building block for a range of amino acids like phenylglycine, which is used as a side chain in various antibiotics. It is involved as a brightener in zinc electroplating processes, and as a food and fragrance additive. It is active intermediate for the manufacture of benzoic acid, odorants and flavoring chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Melting Point -26°C
Boiling Point 178°C to 179°C
Quantity 1000 g
Percent Purity 99+%
Physical Form Liquid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251040 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA10348.0I,A10348.30,A10348.0B
Lot Number10247535 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA10348.0I,A10348.30,A10348.0B
Lot Number10243680 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA10348.0I,A10348.30,A10348.0B
Lot NumberY07F039 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10348.0I,A10348.30,A10348.0B
Lot Number10142256 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10348.0I,A10348.30,A10348.0B
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Benzaldehyde
Product Content Correction
Product Title
Benzaldehyde, 99+%, Thermo Scientific Chemicals >