Learn More
Bel-Art™ Neoprene Sleeved Gloves
Shop All SP Scienceware Products
Click to view available options
Size:
8
11
Description
- One piece
- Made from bonded neoprene
- For glove boxes with 6 in. glove ports
Specifications
Specifications
Color | Black |
Hand | Specific |
Latex Free | No |
Material | Bonded Neoprene |
Product Type | Glove Box Gloves |
Size | 8 |
Product Content Correction
Product Title
Bel-Art™ Neoprene Sleeved Gloves >