Learn More
Melford B-Gentiobiose
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford G36200-1.0
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Substance Name: AmygdaloseMolecular Formula: C12H22O1
Molecular Weight: 342.3
CAS Number: 554-91-6Specifications
554-91-6 | |
C12H22O11 | |
1 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.