Description
- Wide neck wash bottle
- Fitted with color coded one-piece PP closure that maintains it's shape in use
- Designed with integral spout to prevent contamination
Specifications
Specifications
Capacity (Metric) | 500 mL |
Includes | Integral spout |
Closure Color | Red, yellow, blue, green, and orange |
Closure Included | Yes |
Mouth | Wide |
Shape | Round |
Product Type | Wash Bottle |
Chemical Label | Blank |
Closure Material | Polypropylene |
Promotions
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Azlon™ Round Integral Wash Bottle >