Learn More
Azlon™ Round Integral Wash Bottle Product Code.: 15878467
In Stock
to see stock.
Add to basket
Azlon™ Round Integral Wash Bottle
Product Code. 15878467
500 mL
Click to view available options
Capacity (Metric):
500 mL
Product Code. 15878467
Brand: Azlon WGW800P
In Stock
to see stock.
Edge
Add to basket
Add to basket
Azlon™ Round Integral Wash Bottle is molded with an integral spout to reduce contamination caused by the inner tube touching the benchtop during re-filling.
  • Wide neck wash bottle
  • Fitted with color coded one-piece PP closure that maintains it's shape in use
  • Designed with integral spout to prevent contamination
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Includes Integral spout
Closure Color Red, yellow, blue, green, and orange
Closure Included Yes
Mouth Wide
Shape Round
Product Type Wash Bottle
Chemical Label Blank
Closure Material Polypropylene
Product Content Correction
Product Title
Azlon™ Round Integral Wash Bottle >