Learn More
Azlon™ Polyamide Coated Steel Bottle Carrier
Ideal for safely carrying up to six bottles or wash bottles
Brand: Azlon BGS042
89.06 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Manufactured from polyamide coated steel
- Offers better heat and chemical resistance than other coatings
Specifications
Bottle Carrier | |
Polyamide-coated Steel | |
1 | |
74 mm |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%