Learn More
Azlon™ Multi-lingual Round Integral Wash Bottle
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Description
- Multilingual
- Wide neck wash bottle
- Indelibly printed in English, French, German and Spanish
- Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
- Fitted with color coded one-piece PP closure that maintains it's shape in use
- Designed with integral spout to prevent contamination
Specifications
Specifications
Capacity (Metric) | 500 mL |
Includes | Integral spout |
Closure Color | Orange |
Closure Included | Yes |
Mouth | Wide |
Product Type | Wash Bottle |
Chemical Label | Ethanol |
Closure Material | Polypropylene |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
Azlon™ Multi-lingual Round Integral Wash Bottle >