missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 10μL Microvolume Pipette Tips, Non-Filtered
Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes.
Brand: Axygen™ TXL-10-R
This item is not returnable.
View return policy
Description
- Polypropylene tips
- Microvolume
- Long length
- Maxymum Recovery technology for ultra-low retention
- Designed to fit a wide variety of single and multi-channel pipettes
- Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject

Specifications
0.1 to 10 μL | |
Non-sterile | |
USP 3127 | |
Rack | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Polypropylene |
Non-filtered | |
Clear | |
Allows recovery of costly samples with improved precision and accuracy | |
Extended Length, Microvolume | |
Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2 |
Product Suggestions
Customers who viewed this item also viewed
AXY001 Catalogue GENERAL AXYGEN FS
File size: 2.2 MB
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
2Hours
1Minutes
9Seconds
45
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ 10μL Microvolume Pipette Tips, Non-Filtered > Racked; Standard; 0.5-20 μL
Spot an opportunity for improvement?Share a Content Correction