Learn More
WTW™ Armoring with Protective Hood for WTW™ ConOx™ and Tetracon™
The universal arming for the TetraCon™ 325 as well as for the galvanized oxygen sensor CellOx™ 325 provide optimal protection against damage in rough field applications, such as streaming water or bore holes.The special arming A pHLab/K is used to protect SenTix™ pH electrodes with glass shafts when measuring outside the lab.
298.00€ - 344.00€
Specifications
For Use With (Equipment) Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325
Type Armoring
View More Specs
Products 2
Product Code Brand Material For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Material For Use With (Equipment) Price Quantity & Availability  
10613922
View Documents
WTW™
903830
Plastic Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325
298.00€
Each
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
10567831
View Documents
WTW™
903831
Stainless Steel Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description
Description
  • SensoLyt™ 900-P
  • SensoLyt™ ORP 900-P
  • TetraCon™ 925-P
  • FDO™ 925-P
  • VisoTurb™ 900-P
Specifications
Specifications
Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325
Armoring