Learn More
WTW™ Armoring with Protective Hood for WTW™ ConOx™ and Tetracon™
The universal arming for the TetraCon™ 325 as well as for the galvanized oxygen sensor CellOx™ 325 provide optimal protection against damage in rough field applications, such as streaming water or bore holes.The special arming A pHLab/K is used to protect SenTix™ pH electrodes with glass shafts when measuring outside the lab.
Brand:  WTW™ 903831
 View more versions of this product
Product Code. 10567831
Description
Description
  • SensoLyt™ 900-P
  • SensoLyt™ ORP 900-P
  • TetraCon™ 925-P
  • FDO™ 925-P
  • VisoTurb™ 900-P
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325
Armoring
Stainless Steel protective hood
Stainless Steel