Learn More
TNF, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00007124-AP41.1Set
Product Code. 16039868
  • 809.00€ / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-TNF, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TNF, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
TNF
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNF.
DIF/TNF-alpha/TNFA/TNFSF2
Mouse
RUO
7124
Human
Liquid
ELISA
Biotin
TNF
TNF
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNF, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNF, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TNF, Human, Matched Antibody Pair (AP41), Abnova™ > 1 Set