missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin V Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
231.00€ - 462.00€
Specifications
Antigen | Annexin V |
---|---|
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated |
Applications | Western Blot, Immunofluorescence, Gene Knock-Out |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18641381
|
Novus Biologicals
NBP2-92940-0.02ml |
0.02 mL |
231.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18664121
|
Novus Biologicals
NBP2-92940-0.1ml |
0.1 mL |
462.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin V Polyclonal antibody specifically detects Annexin V in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Gene Knock-OutSpecifications
Annexin V | |
Western Blot, Immunofluorescence, Gene Knock-Out | |
Unconjugated | |
Rabbit | |
Apoptosis, Apoptosis Detections | |
PBS with 50% glycerol, pH7.3. | |
308 | |
IgG | |
Affinity purified |
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human Annexin V (NP_001145.1). MAQVLRGTVTDFPGFDERADAETLRKAMKGLGTDEESILTLLTSRSNAQRQEISAAFKTLFGRDLLDDLKSELTGKFEKLIVALMKPSRLYDAYELKHALKGAGTNEKVLTEIIASRTPEELRAIKQVYEEEYGSSLEDDVVGDTSGYYQRMLVVLLQANRDPDAGIDEAQVEQDAQALFQAGELKWGTDEEKFITIFGTRSVSHLRKVFDKYMTISGFQIEETIDRETSGNLEQLLLAVVKSIRSIPAYLAETLYYAMKGAGTDDHTLIRVMVSRSEIDLFNIRKEFRKNFATSLYSMIKGDTSGDYKKALLLLCGEDD | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Annexin V Antibody - BSA Free, Novus Biologicals™