missing translation for 'onlineSavingsMsg'
Learn More
Anion Standard Mixed Concentrate, for Ion Pair Chromatography (for Column Check), Fisher Chemical Product Code.: 10655492

Anion Standard Mixed Concentrate, for Ion Pair Chromatography (for Column Check), Fisher Chemical

Product Code. 10655492
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 10655492

Brand: Fisher Chemical J/4552/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging HDPE plastic bottle
Quantity 100 mL
MDL Number 283866
Physical Form Liquid
Color Colorless
Product Identifier
  • Mixed anion standard, concentrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anion Standard Mixed Concentrate, for Ion Pair Chromatography (for Column Check), Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.