missing translation for 'onlineSavingsMsg'
Learn More
Ammonium sulphate, BAKER ANALYZED™ ACS, J.T. Baker™ Product Code.: 13553840

Ammonium sulphate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 13553840
1 kg
Click to view available options
1 kg
This item is not returnable. View return policy

Product Code. 13553840

Brand: JT Baker 0049.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets ACS Specifications

Specifications

Melting Point 336°C to 339°C
CAS 7783-20-2
pH 5.0 to 6.0 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range min. 99.0%
Molecular Formula H8N2O4S
MDL Number MFCD00003391
Synonym Diammonium sulphate , Sulfuric acid diammonium salt
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name sulfuric acid diamine
Molecular Weight (g/mol) 132.13
Packaging Plastic bottle for solids
Residue after Ignition max. 0.005%
Density 1.78 g/cm³ (20°C)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium sulphate, BAKER ANALYZED™ ACS, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.