missing translation for 'onlineSavingsMsg'
Learn More
Ammonium sulphate, 99.8%, MP Biomedicals™ Product Code.: 11400502

Ammonium sulphate, 99.8%, MP Biomedicals™

Product Code. 11400502 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11400502

Brand: MP Biomedicals 0219140901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7783-20-2
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 0.998
Molecular Formula H8N2O4S
MDL Number MFCD00003391
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name diazanium;sulfate
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
ChEBI CHEBI:62946
Formula Weight 132.1
Chemical Name or Material Ammonium sulphate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium sulphate, 99.8%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.