Learn More
Ammonium (NH4+) (N) Standard for Ion Chromatography, 1000 mg/L in water, Honeywell Fluka™
Brand:  Honeywell Fluka E3002-100ML
Product Code. 16684204
Description
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ammonium (N) Standard for Ion Chromatography
100 mL
1.01 g/cm3
Standards for Ion Chromatography
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Ammonium (NH4+) (N) Standard for Ion Chromatography, 1000 mg/L in water, Honeywell Fluka™ > 100 mL