Learn More
Amidosulfonic acid, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
pH | 1.2 |
Melting Point | ∼205°C (decomposition) |
Density | 2.12 g/mL |
Odor | Odorless |
Quantity | 100 g |
Physical Form | Crystalline |
UN Number | UN2967 |
Merck Index | 148921 |
Solubility Information | Soluble in water. |
Formula Weight | 97.09 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N17L020 | Certificate of Analysis | 26/02/2025 | 044510.36,044510.14,044510.22 |
N26K038 | Certificate of Analysis | 13/09/2024 | 044510.14,044510.36,044510.22 |
S20J043 | Certificate of Analysis | 12/03/2024 | 044510.14,044510.36,044510.22 |
X31I010 | Certificate of Analysis | 09/01/2024 | 044510.14,044510.36,044510.22 |
X07G019 | Certificate of Analysis | 17/01/2023 | 044510.14,044510.36,044510.22 |
Safety and Handling

- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.