missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Western Blotting Sandwiches

Easy-to-use, precut, preassembled PVDF membrane and filter paper sandwiches

165.00€ - 179.00€

Specifications

Length (Metric) 8 cm
Width (Metric) 9 cm
Quantity Pack of 10
Format Sandwich
View More Specs

Products 2
Product Code Brand Length (Metric) Width (Metric) Pore Size Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Pore Size Price Quantity & Availability  
15239884
View Documents
Cytiva
10600119
8 cm 9 cm 0.45 μm
165.00€
Pack of 10
In Stock
to see stock.
Add to basket
 
15249884
View Documents
Cytiva
10600120
8 cm 9 cm 0.2 μm
179.00€
Pack of 10
In Stock
to see stock.
Add to basket
 
Description

Description

  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats

Specifications

Specifications

8 cm
9 cm
Pack of 10
Sandwich
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger||

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.