Learn More
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs
Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
Brand:  Cytiva PA25500
 View more versions of this product
Product Code. 11728278
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
    • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
    • For use in ELISA, immunoprecipitation, and blotting applications.
    • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For the labeling of biological compounds with Cy5.5 bis-functional dye
Cy5.5 bis-Reactive Dye Pack
Labeling 1mg Protein or 30nM Amino groups
Cy5.5 bis-Functional Dye
Each
Five foil packs each containing dried dye to label 1mg of Protein
2°C to 8°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
SDS
Documents
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs > Cy5.5 bis-Reactive Dye Pack