Learn More
Karl Hecht™ Amber Glass Sample Vials with Snap-Cap
Manufactured from amber glass to protect light-sensitive contents.
Brand: Karl Hecht™ 42786020
This item is not returnable.
View return policy
Description
- Flat bottoms
- With rolled rim
- Available in a range of capacities
Specifications
Natural | |
Amber | |
30 mL | |
Snap Cap | |
200/Pk. |
28 mm | |
Glass | |
Polyethylene | |
75 mm | |
Sample Vial |