Learn More
Thermo Scientific™ ALPS Heat Sealer Accessories
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Marke: Thermo Scientific™ AB-0724
269.28 EUR valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Beschreibung
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
Warranty: 1 year
Spezifikation
384-well PCR plate carrier | |
ALPS™ Heat Sealer | |
Carrier |
Sonderangebote
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.