Learn More
Ansell AlphaTec™ 2500 PLUS
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Ultra low linting to help reduce the risk of cross contamination in critical areas
- Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
- Type 3-B liquid tight certified
Specifications
Specifications
Size | X-Large |
Material | Microporous Non-woven Polypropylene Laminate |
Color | White |
Seam Type | Stitched, Taped |
Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
For Use With (Application) | Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments |
Weight (Metric) | 65 gsm |
Style | W25T122 |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
18Hours
18Minutes
32Seconds
8
Product Content Correction
Product Title
Ansell AlphaTec™ 2500 PLUS >